
1,3-Diiodopropane 99% stabilised
:627-31-6
:
:C₃H₆I₂
:MFCD00001097
:295.89 g/mol
:111…113 °C (31 mmHg)
:–20 °C
:2.576 g/cm³
:>110 °C
:210-993-4
:GHS07
- Appearance:Clear light yellow to pink Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6405 to 1.6425 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:Copper