Order Entry
Export
Orders LinkContactUsLinkComponent
 

(±)-Linalool, Sigma-Aldrich®

Irritant
Supplier:   Merck
(±)-Linalool, Sigma-Aldrich®
Supplier:  Merck
Certificates
(±)-Linalool, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • MDL Number:MFCD00008906
  • Molecular Weight:154.25 g/mol
  • Merck Index:13,05517
  • Boiling Pt:199 °C (1013 hPa)
  • Density:0.87 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:75 °C
  • Storage Temperature:Ambient
  • EINECS:201-134-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 52,20 €
    52,20 €
    1 * 5 g
       52,20 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 77,60 €
    77,60 €
    1 * 100 g
       77,60 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 133,00 €
    133,00 €
    1 * 500 g
       133,00 €