(±)-Linalool
Glass bottle for solids, 500 g
Catalog # MOLEM48920082
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
- CAS Number:78-70-6
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00008906
- Molecular Weight:154.25 g/mol
- Merck Index:13,05517
- Boiling Pt:199 °C (1013 hPa)
- Density:0.87 g/cm³ (20 °C)
- Flash Pt:75 °C
- Storage Temperature:Ambient
- EINECS:201-134-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



