Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyvinylpyrrolidone
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
9003-39-8
:
Formula:
(C₆H₉NO)n
MDL Number:
MFCD01076626
Merck Index:
14,7697
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA14315
Videos
Thermo Scientific Acroseal 25 Years
Documentation