Polyvinylpyrrolidone, M.W. K30 (MW ∼ 50.000)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-39-8
:
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626
Melting Pt:225 °C (dec.)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellowish-white Hygroscopic powder
- Infrared spectrum:Conforms
- Water:≤5.0 % (K.F.)
- Sulfated ash:≤0.1 %
- HPLC:≤10 ppm (1-Vinylpyrrolidin-2-one)
- Appearance of solution:(5 % in water at 25°C) Clear
- Viscosity:27 to 32 K-value (1 % w/v in water at 25°C)
- pH:3.0 to 5.0 (5 % in water)
- Nitrogen (N):11.5 to 12.8 % (on anhydrous substance)
- Aldehyde (C2H4O):≤0.05 %
- Hydrazine (N2H4):≤1 ppm
- Peroxides:≤400 ppm