Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyvinylpyrrolidone
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90), Sigma-Aldrich®
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90), Sigma-Aldrich®
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
9003-39-8
:
Formula:
(C₆H₉NO)n
MDL Number:
MFCD01076626
:
M-Clarity™ quality level = MQ200
Storage Temperature:
Ambient