Tween® 80 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9005-65-6
:
MDL Number:MFCD00082107
Melting Pt:100 °C
:M-Clarity™ quality level = MQ300
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:500-019-9