Polyethylene glycol 6,000, Sigma-Aldrich®
Catalog # SIAL03394-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:6000 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:55…62 °C
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779614