Order Entry
Export
ContactUsLinkComponent
Polyethylene glycol 6,000 for biochemistry
Polyethylene glycol 6,000 for biochemistry
Catalog # 442714K
Supplier:  VWR Chemicals
CAS Number:  
undefined
Polyethylene glycol 6,000 for biochemistry
Catalog # 442714K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • IR Spectrum:
    Passes test
  • Average molecular weight:
    5000 - 7000
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Absorbance (260 nm) (0,01 mol/l):
    Max. 0.2
  • Absorbance (280 nm) (0,01 mol/l):
    Max. 0.1

Specification Test Results

About this item

 :
 :6000 g/mol
Boiling Pt:>250 °C
Melting Pt:60…63 °C
Density:1.21 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
 :MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779614