Polyethylene glycol 6,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779614
:6000 g/mol
Boiling Pt:? 250 °C (1013 hPa)
Melting Pt:55…60 °C
Density:1,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-038-2
Specification Test Results
- Appearance:White Powder or flakes
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Hydroxyl value:17 to 20 mg KOH/g