Order Entry
Export
ContactUsLinkComponent
Polyethylene glycol 4,000, Sigma-Aldrich®
Polyethylene glycol 4,000, Sigma-Aldrich®
Catalog # SIAL81242-1KG
Supplier:  Merck
CAS Number:  
Polyethylene glycol 4,000, Sigma-Aldrich®
Catalog # SIAL81242-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :4000 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:53…58 °C
Density:1.13 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
 :MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779612