Order Entry
Export
ContactUsLinkComponent
Polyethylene glycol 300
Polyethylene glycol 300
Certificates
undefined
Polyethylene glycol 300
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4620 to 1.4650 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • Color scale:
    ≤50 APHA
  • Hydroxyl value:
    356 to 394 mg KOH/g

Specification Test Results

Related Information