Order Entry
Export
ContactUsLinkComponent
Polyethylene glycol 20,000, Sigma-Aldrich®
Polyethylene glycol 20,000, Sigma-Aldrich®
Catalog # SIAL95172-250G-F
Supplier:  Merck
CAS Number:  
Polyethylene glycol 20,000, Sigma-Aldrich®
Catalog # SIAL95172-250G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :20000 g/mol
Melting Pt:58…63 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839