Order Entry
Export
ContactUsLinkComponent
Polypropylene glycol, M.W. 2,000
Polypropylene glycol, M.W. 2,000
Certificates
undefined
Polypropylene glycol, M.W. 2,000

Specification Test Results

  • Appearance:
    Clear viscous liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.15 %
  • Color scale:
    ≤50 APHA
  • Viscosity:
    135 to 165 mm²/s (at 40°C)
  • Acid value:
    ≤0.1 mg KOH/g
  • Hydroxyl value:
    53 to 58 mg KOH/g

Specification Test Results

Related Information