
Polyethylene glycol 1,500, Sigma-Aldrich®
Catalog # ROCH10783641001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:40 mL
- Seq:2360
Specifications
About this item
:
:1500 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:44…48 °C
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:500-038-2
:GHS07
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839