Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene glycol 1,500
Polyethylene glycol 1,500
Polyethylene glycol 1,500
:
Thermo Fisher Scientific
Polyethylene glycol 1,500
:
Thermo Fisher Scientific
:
25322-68-3
:
:
H(OCH₂CH₂)
n
OH
:
MFCD00081839
:
1500 g/mol
:
14,7568
:
∼250 °C (1013 hPa)
:
44…48 °C
:
1.13 g/cm³ (20 °C)
:
260 °C
:
500-038-2
:
GHS07
:
Ambient
msds_ALFAA16241
Thermo Scientific Acroseal 25 Years