
Polyethylene glycol 1,500
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779609
:1500 g/mol
Boiling Pt:? 250 °C (1013 hPa)
Melting Pt:44…48 °C
Density:1,13 g/cm³ (20 °C)
Flash Pt:>250 °C
EINECS:500-038-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Waxy flakes
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- Hydroxyl value:70 to 80 mg KOH/g