Order Entry
Export
ContactUsLinkComponent
Bovine serum albumin (BSA), Fraction V
Bovine serum albumin (BSA), Fraction V
Catalog # ROCH10735086001
Supplier:  ROCHE
CAS Number:  
Bovine serum albumin (BSA), Fraction V
Catalog # ROCH10735086001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • CAS No.:
    90604-29-8
    9048-46-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin

Specifications

About this item