Order Entry
Export
Orders LinkContactUsLinkComponent
Gelatin

Gelatin

Supplier:   Roth Carl
Gelatin
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ROTH4308.1
  • ROTH4308.2
  • Protein/peptide name:
  • Gelatin
  • Gelatin
  • Protein synonyms:
  • Gelatine
  • Gelatine
  • CAS No.:
  • 9000-70-8
  • 9000-70-8
  • Storage conditions:
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 55,60 €
    55,60 €
    1 * 1 kg
       55,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    ROTH4308.1
    Protein/peptide name
    Gelatin
    Protein synonyms
    Gelatine
    CAS No.
    9000-70-8
    Storage conditions
    Ambient
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 226,00 €
    226,00 €
    1 * 5 kg
       226,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ROTH4308.2
    Protein/peptide name
    Gelatin
    Protein synonyms
    Gelatine
    CAS No.
    9000-70-8
    Storage conditions
    Ambient