0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Enzymes
Xanthine oxidase
Xanthine oxidase
ROCH10110434001
:
ROCHE
:
Xanthine oxidase
ROCH10110434001
:
ROCHE
:
Pk:
20 AU
CAS No.:
9002-17-9
Seq:
01
Enzyme name:
Xanthine oxidase