Order Entry
Export
Orders LinkContactUsLinkComponent
Agarose

Agarose

Supplier:   Roth Carl
Agarose
Supplier:  Roth Carl
Certificates
Agarose

3 Options Available Below

Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 29,60 €
    29,60 €
    1 * 10 g
       29,60 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 161,00 €
    161,00 €
    1 * 100 g
       161,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 110,00 €
    1 110,00 €
    1 * 1 kg
       1 110,00 €