Agar, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Agar Agar
- CAS Number:9002-18-0
- Formula:C₁₂nH₁₈nOn₉n
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:232-658-1
2 Options Available Below
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- Formula:C₁₂nH₁₈nOn₉n
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:232-658-1



