Order Entry
Export
Orders LinkContactUsLinkComponent
 

Agar, Sigma-Aldrich®

Supplier:   Merck
Agar, Sigma-Aldrich®
Supplier:  Merck
Certificates
Agar, Sigma-Aldrich®
Agar, Sigma-Aldrich®

5 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:C₁₂nH₁₈nOn₉n
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:232-658-1

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 80,10 €
    80,10 €
    1 * 100 g
       80,10 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 143,00 €
    143,00 €
    1 * 250 g
       143,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 236,00 €
    236,00 €
    1 * 500 g
       236,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 426,00 €
    426,00 €
    1 * 1 kg
       426,00 €

  • Glass bottle for solids
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 1 710,00 €
    1 710,00 €
    1 * 5 kg
       1 710,00 €