Order Entry
Export
Orders LinkContactUsLinkComponent
Agar, Sigma-Aldrich®

Agar, Sigma-Aldrich®

Supplier:   Merck
Agar, Sigma-Aldrich®
Supplier:  Merck
Certificates
Agar, Sigma-Aldrich®
Agar, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:C₁₂nH₁₈nOn₉n
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 380,00 €
    380,00 €
    1 * 500 g
       380,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 455,00 €
    455,00 €
    1 * 1 kg
       455,00 €