Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cellulose

20 kg
Catalog # MSPPVIVAPUR102
Cellulose
Catalog # MSPPVIVAPUR102
CAS Number:  9004-34-6
Cellulose

20 kg

CAS Number:  9004-34-6
Price
Inquire For Price
Log in for availability
1 * 20 kg
Product Details & Documents
Synonyms:  Microcrystallinecellulose
  • CAS Number:9004-34-6
  • Formula:(C₁₂H₂₂O₁₁)n
  • MDL Number:MFCD00081512
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,01977
  • Melting Pt:260…270 °C
  • Density:1.27…1.6 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-674-9
Specifications

Specifications

  • Pk:
    20 kg

Specifications