Cellulose, Sigma-Aldrich®
Catalog # SIAL11363-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:342.3 g/mol
Merck Index:13,01977
Melting Pt:260…270 °C
Density:1.27…1.6 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:232-674-9
CAS Number:9004-34-6
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512