Order Entry
Export
Orders LinkContactUsLinkComponent
Streptomycin sulphate

Streptomycin sulphate

Health
Irritant
Supplier:   Roth Carl
Streptomycin sulphate
Supplier:  Roth Carl
Certificates
Streptomycin sulphate

3 Options Available Below

Product Details & Documents
  • CAS Number:3810-74-0
  • Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
  • MDL Number:MFCD00037023
  • Molecular Weight:1457.4 g/mol
  • Merck Index:12,08983
  • Melting Pt:250 °C
  • Storage Temperature:Refrigerator
  • EINECS:223-286-0
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 24,90 €
    24,90 €
    1 * 10 g
       24,90 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 41,50 €
    41,50 €
    1 * 25 g
       41,50 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 67,00 €
    67,00 €
    1 * 50 g
       67,00 €