Order Entry
Export
Orders LinkContactUsLinkComponent
Streptomycin sulphate

Streptomycin sulphate

Glass bottle for solids, 100 g
Catalog # MOLE17865212-100G
Health
Irritant
Supplier:   Molekula
Streptomycin sulphate
Catalog # MOLE17865212-100G
Supplier:  Molekula
CAS Number:  3810-74-0
Streptomycin sulphate

Glass bottle for solids, 100 g

CAS Number:  3810-74-0
Price
145,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:3810-74-0
  • Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
  • MDL Number:MFCD00037023
  • Molecular Weight:1457.4 g/mol
  • Merck Index:12,08983
  • Melting Pt:250 °C
  • Storage Temperature:Refrigerator
  • EINECS:223-286-0
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications