
Gentamicin sulphate
:1405-41-0
:
:MFCD00270181
:13,04403
:218…237 °C
:215-778-9
:GHS08
:Ambient
- Synonyms:Garamycin; Gentamicin sulphate salt; Gentiomycin C
- Form:White powder
- Storage:Store at +4 °C
- Loss on Drying:≤18,0 %
- Optical Rotation:107° to +121° (10 mg/ml, water)
- pH:3,5 - 5,5 (1 in 25, in water)
- Potency:≥590 µg/mg