Order Entry
Export
ContactUsLinkComponent
Tween® 20 (Polysorbate) 10% in aqueous solution, Sigma-Aldrich®
Tween® 20 (Polysorbate) 10% in aqueous solution, Sigma-Aldrich®
Catalog # ROCH11332465001
Supplier:  ROCHE
CAS Number:  
Tween® 20 (Polysorbate) 10% in aqueous solution, Sigma-Aldrich®
Catalog # ROCH11332465001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :1227.53 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986