Order Entry
Export
ContactUsLinkComponent
 
Corrosive
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, Sigma-Aldrich®
Catalog # ROCH11332481001
Supplier:  ROCHE
CAS Number:  
Corrosive
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, Sigma-Aldrich®
Catalog # ROCH11332481001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
Boiling Pt:∼100 °C (1013 hPa)
Density:∼1.01 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :GHS05
Storage Temperature:Ambient
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505