L(+)-Arabinose
Glass bottle for solids, 100 g
Catalog # MOLE12030733-100G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Pectinose, Pectin Sugar
- CAS Number:5328-37-0
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00067709
- Molecular Weight:150.13 g/mol
- Boiling Pt:331 °C (1013 hPa)
- Melting Pt:158…160 °C
- Density:1.585 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-214-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



