Order Entry
Export
ContactUsLinkComponent
D-(+)-Mannose
D-(+)-Mannose
Catalog # EHERC14752300
Supplier:  EHRENSTORFER
CAS Number:  
D-(+)-Mannose
Catalog # EHERC14752300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
 :
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:222-392-4
CAS Number:3458-28-4
 :MFCD00799233
Formula:C₆H₁₂O₆
MDL Number:MFCD00064122
Boiling Pt:376
Melting Pt:133
Density:1.539 g/cm³ (20 °C)
Merck Index:13,05772