Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Glucose

D-(+)-Glucose

5 kg
Catalog # MOLE13002238
Supplier:   Molekula
D-(+)-Glucose
Catalog # MOLE13002238
Supplier:  Molekula
CAS Number:  50-99-7
D-(+)-Glucose
D-(+)-Glucose

5 kg

CAS Number:  50-99-7
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,04459
  • Melting Pt:146 °C
  • Density:1.54 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-075-1
Specifications

Specifications

  • Pk:
    5 kg

Specifications