Order Entry
Export
ContactUsLinkComponent
 
D-(+)-Galactose
Catalog # EHERC13996500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
D-(+)-Galactose
Catalog # EHERC13996500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :180.16 g/mol
Merck Index:13,04356
Boiling Pt:527 °C (1013 hPa)
Melting Pt:163…169 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-416-4
CAS Number:59-23-4
Formula:C₆H₁₂O₆
MDL Number:MFCD00151230