Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Sucrose, Millipore®

D-(+)-Sucrose, Millipore®

Supplier:   Merck
D-(+)-Sucrose, Millipore®
Supplier:  Merck
Certificates
D-(+)-Sucrose, Millipore®
D-(+)-Sucrose, Millipore®

3 Options Available Below

Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-334-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 25,20 €
    25,20 €
    1 * 250 g
       25,20 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 70,80 €
    70,80 €
    1 * 1 kg
       70,80 €

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 263,00 €
    263,00 €
    1 * 5 kg
       263,00 €