D-(+)-Sucrose, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57-50-1
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
Merck Index:13,08966
Melting Pt:169…170 °C
Density:1.29 g/cm³ (20 °C)
:342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9