D-(+)-Sucrose
Plastic bottle for solids, 5 kg
Catalog # MOLEM17975892
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
- CAS Number:57-50-1
- Formula:C₁₂H₂₂O₁₁
- MDL Number:MFCD00006626
- Molecular Weight:342.3 g/mol
- Merck Index:13,08966
- Melting Pt:169…170 °C
- Density:1.29 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-334-9
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 kg



