Order Entry
Export
ContactUsLinkComponent
 
D-(+)-Sucrose
Catalog # EHERC16901100
Supplier:  EHRENSTORFER
CAS Number:  
undefined
D-(+)-Sucrose
Catalog # EHERC16901100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
Merck Index:13,08966
Melting Pt:169…170 °C
Density:1.29 g/cm³ (20 °C)
 :342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9
CAS Number:57-50-1
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626