D-(+)-Raffinose pentahydrate, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:17629-30-0
:
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590
:594.52 g/mol
Merck Index:12,08279
Melting Pt:80 °C
Density:1.516 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:208-146-9