Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Raffinose pentahydrate

D-(+)-Raffinose pentahydrate

Supplier:   Roth Carl
D-(+)-Raffinose pentahydrate
Supplier:  Roth Carl
Certificates
D-(+)-Raffinose pentahydrate

2 Options Available Below

Product Details & Documents
Synonyms:  D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate
  • CAS Number:17629-30-0
  • Formula:C₁₈H₃₂O₁₆·5H₂O
  • MDL Number:MFCD00071590
  • Molecular Weight:594.52 g/mol
  • Merck Index:12,08279
  • Melting Pt:80 °C
  • Density:1.516 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:208-146-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 27,60 €
    27,60 €
    1 * 10 g
       27,60 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 88,80 €
    88,80 €
    1 * 50 g
       88,80 €