Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Glucose monohydrate
D-(+)-Glucose monohydrate, Millipore®
D-(+)-Glucose monohydrate, Millipore®
:
Merck
D-(+)-Glucose monohydrate, Millipore®
:
Merck
:
14431-43-7
:
5996-10-1
:
C₆H₁₂O₆·H₂O
:
MFCD00149450
:
198.17 g/mol
:
83 °C
:
M-Clarity™ quality level = MQ200
:
Ambient
:
200-075-1