Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Trehalose dihydrate

D-(+)-Trehalose dihydrate

Supplier:   Roth Carl
D-(+)-Trehalose dihydrate
Supplier:  Roth Carl
Certificates
D-(+)-Trehalose dihydrate
D-(+)-Trehalose dihydrate

3 Options Available Below

Product Details & Documents
Synonyms:  Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
  • CAS Number:6138-23-4
  • Formula:C₁₂H₂₂O₁₁·2H₂O
  • MDL Number:MFCD00071594
  • Molecular Weight:378.33 g/mol
  • Melting Pt:97…99 °C
  • Storage Temperature:Ambient
  • EINECS:202-739-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 17,30 €
    17,30 €
    1 * 5 g
       17,30 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 46,20 €
    46,20 €
    1 * 25 g
       46,20 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 352,00 €
    352,00 €
    1 * 250 g
       352,00 €