Order Entry
Export
ContactUsLinkComponent
D-(+)-Trehalose dihydrate
D-(+)-Trehalose dihydrate
Catalog # EHERC17607500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
D-(+)-Trehalose dihydrate
Catalog # EHERC17607500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :378.33 g/mol
Melting Pt:97…99 °C
Storage Temperature:Ambient
EINECS:202-739-6
CAS Number:6138-23-4
Formula:C₁₂H₂₂O₁₁·2H₂O
MDL Number:MFCD00071594