D-(+)-Trehalose dihydrate
Glass bottle for solids, 100 g
Catalog # BMOL54688-2
Supplier: BIOMOL RESEARCH LABORATORIES
D-(+)-Trehalose dihydrate
Catalog # BMOL54688-2
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number: 6138-23-4
Jump to:
Product Details & Documents
Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
- CAS Number:6138-23-4
- Formula:C₁₂H₂₂O₁₁·2H₂O
- MDL Number:MFCD00071594
- Molecular Weight:378.33 g/mol
- Melting Pt:97…99 °C
- Storage Temperature:Ambient
- EINECS:202-739-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



