Benzalkonium chloride
Glass bottle for solids, 100 g
Catalog # MOLEM84982516
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
- CAS Number:63449-41-2
- Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
- MDL Number:MFCD00145757
- Melting Pt:29…34 °C
- Density:0.98 g/cm³ (20 °C)
- EINECS:264-151-6
- UN:2923
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



