
Fullerene C60 99.9%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99685-96-8
:
Formula:C₆₀
MDL Number:MFCD00151408
Merck Index:13,01447
Melting Pt:? 280 °C
Density:~ 1,65 g/cm³ (20 °C)
ADR:4.1,III
:720.66 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown to black Powder
- Infrared spectrum:Conforms
- HPLC:≥99.85 %