



Petroleum spirit, 60…95 °C, AnalaR NORMAPUR® analytical reagent
Catalog # 1991.1000
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Heavy oils and fats:Passes test
- Acidity:Max. 0.0001 meq/g
- Evaporation residue:Max. 10 ppm
- Sulphur compounds (as S):Max. 50 ppm
- Water:Max. 0.02 %
Specification Test Results
About this item
:
Boiling Pt:60…95 °C (1013 hPa)
Melting Pt:≤–40 °C
Density:0.67 g/cm³ (20 °C)
Flash Pt:–21 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849