



Petroleum spirit, 60…95 °C, extra pure
:64742-49-0
:8032-32-4
:MFCD00081849
:60…95 °C (760 mmHg)
:–40 °C
:0.7 g/cm³
:–20 °C
:Ambient
:265-151-9
:3295
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3880 to 1.3920 (20°C, 589 nm)
- Boiling point:60 to 95°C
- Density:(15°C) 0.690 to 0.710 g/ml
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- n-Hexane:≤2 %
- Benzene:≤35 ppm
- Aromatics:≤0.01 %