



Petroleum spirit, 35…60 °C, extra pure
Catalog # ACRO413140010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3660 to 1.3710 (20°C, 589 nm)
- Boiling point:35 to 60°C
- Density:(15°C) 0.649 to 0.659 g/ml
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- n-Hexane:≤3 %
- Benzene:≤55 ppm
- Aromatics:≤0.01 %
Specification Test Results
About this item
:
Boiling Pt:35…60 °C (1013 hPa)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS08
:GHS09
:GHS02
:GHS07
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849